EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Analysis of beers from an 1840s' shipwreck.
PMID 25664918
Abstract
Two bottles of beer from an about 170-year-old shipwreck (M1 Fö 403.3) near the Åland Islands in the Baltic Sea were analyzed. Hop components and their degradation compounds showed that the bottles contained two different beers, one more strongly hopped than the other. The hops used contained higher levels of β-acids than modern varieties and were added before the worts were boiled, converting α-acids to iso-α-acids and β-acids to hulupones. High levels of organic acids, carbonyl compounds, and glucose indicated extensive bacterial and enzyme activity during aging. However, concentrations of yeast-derived flavor compounds were similar to those of modern beers, except that 3-methylbutyl acetate was unusually low in both beers and 2-phenylethanol and possibly 2-phenylethyl acetate were unusually high in one beer. Concentrations of phenolic compounds were similar to those in modern lagers and ales.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
45840
Acetic anhydride
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
32900
Benalaxyl-M, PESTANAL®, analytical standard
C20H23NO3
B6768
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
H3BO3
B7901
Boric acid, for electrophoresis, ≥99.5%
H3BO3
B2645
Boric acid, tablet, 1 g boric acid per tablet
H3BO3
202878
Boric acid, 99.999% trace metals basis
H3BO3
339067
Boric acid, 99.97% trace metals basis
H3BO3
B7660
Boric acid, BioXtra, ≥99.5%
H3BO3
B0394
Boric acid, ACS reagent, ≥99.5%
H3BO3
B0252
Boric acid, ReagentPlus®, ≥99.5%
H3BO3
15663
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
H3BO3
31146
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
H3BO3
11607
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
H3BO3
426148
Boric acid-11B, ≥99 atom % 11B
H311BO3
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
09-1833
Diethyl ether, for residue analysis, JIS 5000
C4H10O
09-1855
Diethyl ether
C4H10O
N7502
DL-Norvaline
C5H11NO2
M0789
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥97.0% (GC)
C5H3F6NO2
65943
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥99.0% (GC)
C5H3F6NO2
84727
Sulfuric acid, for the determination of nitrogen, ≥97.5%
H2O4S
68279
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
H2O4S