EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Protective effect of panax notoginseng saponins on acute ethanol-induced liver injury is associated with ameliorating hepatic lipid accumulation and reducing ethanol-mediated oxidative stress.
PMID 25665731