EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Simultaneous GC-MS determination of eight phthalates in total and migrated portions of plasticized polymeric toys and childcare articles.
PMID 25667041
Abstract
A gas chromatography/mass spectrometric method was developed for the simultaneous determination of eight phthalic acid esters (PAEs) in toys and child care articles that are made of plasticized plastic. The novel method was used to determine the total concentration of the PAEs in addition to the migrated PAEs values into artificial saliva, under conditions that simulate real life situations. The extraction method, which was developed for the first time to determine the total concentration of PAEs, utilized a novel optimization of four parameters involving the solvent, time, temperature and weight of sample. The PAEs were extracted with tetrahydrofuran, as extraction the solvent, and using the ultrasonic water bath shaker for 30min, at room temperature. Another extraction method was developed to determine the migrated PAEs into artificial saliva at pH 6.2 and 37°C, implementing a liquid-liquid extraction with chloroform. Both methods were subjected to validation steps in terms of linearity, precision, accuracy and recovery, which ensured that all obtained results were well within the norms of acceptable limits and specifications. The analytes were separated at the following retention times: 4.99, 5.21, 5.31, 6.63, 7.41, 9.05min for di-"isobutyl" phthalate (DIBP), dibutyl phthalate (DBP), bis(2-methoxyethyl)phthalate (DMEP), benzyl butyl phthalate (BBP), dibutyl phthalate (DEHP) and di-n-octyl phthalate (DnOP), respectively. The chromatographic peaks corresponding to di-"isononyl" phthalate (DINP) and di-"isononyl" phthalate (DIDP), were separated, using the extracted ion chromatogram (EIC) mode within the time ranges of 8.05-12.10min for DINP and 8.50-14.50min for DIDP. The instrument detection limits for DIBP, DBP, DMEP, BBP, DEHP, DnOP, DINP and DIDP were determined at 0.100, 0.100, 0.045, 0.035, 0.015, 0.370, 0.320, 0.260μg/ml, respectively. The calibration curve working ranges were determined at 0.5-25μg/ml for DIBP, DBP, DMEP, BBP and DEHP, 2-100μg/ml for DnOP and 5-100μg/ml for DINP and DIDP. The coefficients of variation (precision) for the total PAEs method were in the range 0.55-12.10% and for the migration method were in the range 0.72-7.70%. Recovery values for total PAEs and for the migrated PAEs were ranged within 90.6-111.7% and 86.3-117.2%; respectively. Both developed methods were used to survey the Jordanian market, where twenty seven samples were collected and tested for plastic type, total PAEs and migrated PAEs. Seventeen of those samples were found to be polyvinyl chloride (PVC), and all of them containing total PAEs having concentrations in excess of the allowed limit of 0.1% (w/w). The rest of the samples were non-PVC and only one of them containing PAEs, but below the allowed limit. In contrast, all the migration results were found to be well within the allowed limits. It was also found that as the PAEs total concentration increase, the corresponding migrated value increase, while the opposite trend holds for in the PAEs molecular weight.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6630
Benzyl benzoate, ReagentPlus®, ≥99.0%
C14H12O2
B9550
Benzyl benzoate, meets USP testing specifications
C14H12O2
1062008
Benzyl benzoate, United States Pharmacopeia (USP) Reference Standard
C14H12O2
68183
Benzyl benzoate, analytical standard
C14H12O2
12392
Benzyl benzoate, tested according to Ph Eur
C14H12O2
W213802
Benzyl benzoate, ≥99%, FCC, FG
C14H12O2
W213810
Benzyl benzoate, natural, ≥99%, FCC, FG
C14H12O2
PHR1213
Benzyl benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
C14H12O2
42438
Benzyl butyl phthalate, certified reference material, TraceCERT®
C19H20O4
36927
Benzyl butyl phthalate, analytical standard
C19H20O4
308501
Benzyl butyl phthalate, 98%
C19H20O4
442503
Benzyl butyl phthalate, analytical standard
C19H20O4
67261
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®
C24H38O4
36735
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
C24H38O4
80030
Bis(2-ethylhexyl) phthalate, Selectophore
C24H38O4
88173
Di-n-octyl phthalate, certified reference material, TraceCERT®
C24H38O4
31301
Di-n-octyl phthalate, analytical standard
C24H38O4
80153
Di-n-octyl phthalate, ≥98.0% (GC)
C24H38O4
48560-U
Di-n-octyl phthalate, analytical standard
C24H38O4
43540
Diisobutyl phthalate, certified reference material, TraceCERT®
C16H22O4
152641
Diisobutyl phthalate, 99%
C16H22O4
D201154
Dioctyl phthalate, ≥99.5%
C24H38O4
P2155001
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
C24H38O4