EMAIL THIS PAGE TO A FRIEND
Infection and Immunity
Antibodies against hemolysin and cytotoxic necrotizing factor type 1 (CNF1) reduce bladder inflammation in a mouse model of urinary tract infection with toxigenic uropathogenic Escherichia coli.
PMID 25667267
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1278
Galantide, ≥90% (HPLC)
C104H151N25O26S
G0355000
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
C5H9NO4