EMAIL THIS PAGE TO A FRIEND
Anticancer research
In vitro evaluation of 3-arylcoumarin derivatives in A549 cell line.
PMID 25667442
Abstract
Coumarins are naturally-occurring compounds with diverse and interesting biological activities. In the present study, we evaluated the in vitro cytotoxic effect of 8-(acetyloxy)-3-[4-(acetyloxy)phenyl]-2-oxo-2H-chromen-7-yl acetate (6); 8-(acetyloxy)-3-(4-methanesulfonyl phenyl)-2-oxo-2H-chromen-7-yl acetate (7); 4-(2-oxo-2H-chromen-3-yl)phenyl acetate (8); 3-(4-methanesulfonylphenyl)-2H-chromen-2-one (9); 4-(4-methyl-2-oxo-2H-chromen-3-yl)phenyl acetate (10); 3-(4-methanesulfonylphenyl)-4-methyl-2H-chromen-2-one (11); 8-(acetyloxy)-3-[4-(acetyloxy)phenyl]-4-methyl-2-oxo-2H-chromen-7-yl acetate (12); and 5-(acetyloxy)-3-[4-(acetyloxy) phenyl]-2-oxo-2H-chromen-7-yl acetate (13) in human lung (A549) cancer and normal lung (MRC-9) cell lines at different concentrations for 48 h using crystal violet dye binding assay. The cytotoxic effect of these coumarin derivatives were compared to the standard drug, docetaxel. Furthermore, the effect of the most active compound on the cell-cycle using propidium iodide, mitochondrial membrane potential (MMP) using tetramethyl rhodamine methyl ester (rhodamine-123) and reactive oxygen species (ROS) production using 2',7'-dichlorofluorescin diacetate (PCFDA) were also evaluated. Compound 7: had the greatest cytotoxic effect (cytotoxic concentration, CC50=24 μM) and selectivity (MRC-9; CC50>100 μM; inactive) in the A549 cell line, and caused cells to arrest in the S phase of the cell cycle, loss of MMP and increased ROS production in a concentration-dependent manner. These findings suggest that compound 7: could serve as a new lead for the development of novel synthetic compounds with enhanced anticancer activity.