EMAIL THIS PAGE TO A FRIEND
Anticancer research
Vitamin D inhibits ovarian cancer cell line proliferation in combination with celecoxib and suppresses cyclooxygenase-2 expression.
PMID 25667511
Abstract
Vitamin D exhibits multiple anti-proliferative and pro-differentiating actions. Prostaglandin-(PG)E2 is a tumor-promoting tissue hormone anabolized by cyclooxygenase-2 (COX-2). Recently, a link between the PG and vitamin D metabolism was reported. The influence of calcitriol and celecoxib on the proliferation of ovarian cancer cell lines was measured and the impact of calcitriol on the protein and mRNA expression of COX-2 was quantified by western blot and qRT-PCR, respectively. After COX-2 induction with interleukin (IL)-1β, 10 μM celecoxib did not significantly inhibit the proliferation of OVCAR-3 cells, whereas calcitriol showed such an effect; however, the combination of the two substances had an additive influence. After induction by IL-1β, calcitriol inhibited the COX-2 protein, as well as its mRNA expression significantly in OVCAR-3 and SKOV-3 cells. These data suggest a correlation between PG and vitamin D metabolism in their anti-tumorigenic activity in ovarian carcinomas.