EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Naringenin suppresses K562 human leukemia cell proliferation and ameliorates Adriamycin-induced oxidative damage in polymorphonuclear leukocytes.
PMID 25667616
Abstract
Treatments for leukemia remain unsatisfactory. Conventional chemotherapy agents that aim to kill tumor cells may also damage normal cells and thus result in severe side-effects. Naringenin, a natural polyphenolic compound with antioxidant effects, has been revealed to have significant antitumor effects with low toxicity in preliminary studies. Thus, it is considered as one of the most promising flavonoids in the treatment of leukemia. In the present study, the effects of naringenin on the K562 human leukemia cell line and the underlying mechanisms were explored
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52186
(±)-Naringenin, analytical standard
C15H12O5