EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Determining trace amounts and the origin of formaldehyde impurity in Neisseria meningitidis A/C/Y/W-135-DT conjugate vaccine formulated in isotonic aqueous 1? PBS by improved C18-UPLC method.
PMID 25668795