EMAIL THIS PAGE TO A FRIEND
Theriogenology
Serum tryptophan and its metabolites in female dogs undergoing ovariohysterectomy as treatment of pyometra or as elective spay surgery.
PMID 25670154
Abstract
This study compares serum concentrations of tryptophan (TRP), kynurenine (KYN), kynurenic acid (KYNA), and indoleamine 2,3-dioxygenase (IDO) activity in healthy bitches and bitches with bacterial uterine infection (pyometra). The effects of surgery were also assessed by measuring these variables in both groups of dogs before and after ovariohysterectomy. Presurgery, mean (±standard deviation) TRP, KYN, and KYNA concentrations and IDO activity were 68.44 ± 21.77, 2.00 ± 0.33, 112.11 ± 111.91 μmol/L, and 29.22 ± 10.10, respectively, in the healthy dogs; and 40.16 ± 12.11, 8.27 ± 3.94, 411.11 ± 199.60 μmol/L, and 205.92 ± 154.20, respectively, in the dogs with pyometra. Tryptophan and KYN levels had normalized on suture removal (10 days after surgery) though IDO activity and KYNA concentrations remained elevated during the postoperative period compared with presurgery values in both study groups. Our results suggest that KYNA concentrations and IDO activity could be useful indicators of the inflammation induced by pyometra and could be also used to monitor recovery following ovariohysterectomy in both healthy dogs and dogs with pyometra.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2610000
Tryptophan, European Pharmacopoeia (EP) Reference Standard
C11H12N2O2