EMAIL THIS PAGE TO A FRIEND
Photochemical & Photobiological Sciences : Official Journal of the European Photochemistry Association and the European Society for Photobiology
A safety study of a novel photosensitizer, sinoporphyrin sodium, for photodynamic therapy in Beagle dogs.
PMID 25671327