EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Rapid and sensitive detection of the food allergen glycinin in powdered milk using a lateral flow colloidal gold immunoassay strip test.
PMID 25671495