EMAIL THIS PAGE TO A FRIEND
International journal of oncology
CCNJ detected by triple combination array analysis as a tumor-related gene of hepatocellular carcinoma.
PMID 25672416
Abstract
Hepatocellular carcinoma (HCC) has a high likelihood of recurrence and a poor prognosis. To detect cancer-related genes of HCC, we developed a new technique: triple combination array analysis, consisting of a methylation array, a gene expression array and a single nucleotide polymorphism array. A surgical specimen obtained from a 68-year-old female HCC patient was analyzed using triple combination array, which identified cyclin J (CCNJ) as a candidate cancer-related gene of HCC. Subsequently, samples from 85 HCC patients were evaluated for CCNJ promoter hypermethylation and expression status using methylation-specific PCR (MSP) and quantitative reverse transcriptase RT-PCR, respectively. CCNJ was found to be hypermethylated (methylation value, 0.906; range, 0-1.0) in cancer tissue, compared with adjacent non-cancerous tissue (0.112) using a methylation array. MSP revealed that CCNJ was hypermethylated in 67 (78.8%) of the tumor samples. CCNJ expression was significantly decreased in cases with hypermethylation (P<0.0001). Furthermore, cases with both promoter hypermethylation and decreased expression of CCNJ in the tumor tissue had a worse overall survival than the other cases (P=0.0383). In conclusion, our results indicated that CCNJ could be a novel prognostic marker of HCC, and this study indicated that triple combination array analysis was effective in detecting new tumor-related genes and their mechanisms.