EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Medicine
Resveratrol exerts growth inhibitory effects on human SZ95 sebocytes through the inactivation of the PI3-K/Akt pathway.
PMID 25672876