EMAIL THIS PAGE TO A FRIEND
International Journal of Oncology
The anthraquinone derivative Emodin inhibits angiogenesis and metastasis through downregulating Runx2 activity in breast cancer.
PMID 25673059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7881
Emodin, from Frangula bark, ≥90% (HPLC)
C15H10O5
S6317
L-Sulforaphane, ≥95% (HPLC), oil
C6H11NOS2