EMAIL THIS PAGE TO A FRIEND
Journal of drug targeting
Gene silencing of Rac1 with RNA interference mediated by ultrasound and microbubbles in human LoVo cells: evaluation of cell invasion inhibition and metastatic.
PMID 25673262
Abstract
The objective of this study was to assess the change of cytoskeleton and cell cycle in LoVo (human colorectal cancer) cell via gene silencing of Rac1 with RNA interference mediated by microbubble (SonoVue) and ultrasound (US). The compound of plasmid Rac1-shRNA, LoVo cells, and SonoVue was exposed to US (1 MHz, 2 W/cm(2), 5 min). The expression of Rac1 mRNA and Rac1 protein was detected by RT-PCR and Western blot. Cytoskeleton was taken by confocal microscope in a random fashion. Cell invasion was assayed using modified Boyden chambers, and cell cycle and apoptosis were analyzed by flow cytometry in LoVo cells. Rac1 gene is overexpressed in human colorectal cancer cells, gene silencing of Rac1 with RNA interference mediated by microbubble and US strongly inhibited lamellipodia formation, cell invasion, and delayed cell cycle, as well as enhanced cell apoptosis of LoVo cells in vitro. Silencing Rac1 gene mediated by microbubble and US may become a new treatment option for the inhibition of the invasion and metastasis of colorectal cancer cells.