EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Inflammatory monocytes recruited to the liver within 24 hours after virus-induced inflammation resemble Kupffer cells but are functionally distinct.
PMID 25673700