EMAIL THIS PAGE TO A FRIEND
Journal of Cell Science
Nek2A destruction marks APC/C activation at the prophase-to-prometaphase transition by spindle-checkpoint-restricted Cdc20.
PMID 25673878
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3904
Reversine, ≥98% (HPLC), solid
C21H27N7O