EMAIL THIS PAGE TO A FRIEND
Cell Death & Disease
Mitochondrial Lon regulates apoptosis through the association with Hsp60-mtHsp70 complex.
PMID 25675302
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45533
3-Indoleacetic acid, PESTANAL®, analytical standard
C10H9NO2