EMAIL THIS PAGE TO A FRIEND
Chemistry and Biodiversity
Antibacterial activity of enrofloxacin and ciprofloxacin derivatives of ?-octaarginine.
PMID 25676502
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7000
Deoxycorticosterone acetate
C23H32O4
91238
Diethyl ether, analytical standard
C4H10O