EMAIL THIS PAGE TO A FRIEND
OncoTargets and therapy
Erratum: Disruption of human papillomavirus 16 E6 gene by clustered regularly interspaced short palindromic repeat/Cas system in human cervical cancer cells [Corrigendum].
PMID 25676591
Abstract
[This corrects the article on p. 37 in vol. 8, PMID: 25565864.].