EMAIL THIS PAGE TO A FRIEND
The Journal of Pharmacy and Pharmacology
Correlating liposomal adjuvant characteristics to in-vivo cell-mediated immunity using a novel Mycobacterium tuberculosis fusion protein: a multivariate analysis study.
PMID 25677402