EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Determination of GDC-0980 (apitolisib), a small molecule dual phosphatidylinositide 3-kinase/mammalian target of rapamycin inhibitor in dog plasma by LC-MS/MS to support a GLP toxicology study.
PMID 25677784
Abstract
An LC-MS/MS method for the determination of GDC-0980 (apitolisib) concentrations in dog plasma has been developed and validated for the first time to support pre-clinical drug development. Following protein precipitation with acetonitrile, the resulting samples were analyzed using reverse-phase chromatography on a Metasil AQ column. The mass analysis was performed on a triple quadruple mass spectrometer coupled with an electrospray interface in positive ionization mode. The selected reaction monitoring transitions monitored were m/z 499.3 → 341.1 for GDC-0980 and m/z 507.3 → 341.1 for IS. The method was validated over the calibration curve range 0.250-250 ng/mL with linear regression and 1/x(2) weighting. Relative standard deviation (RSD) ranged from 0.0 to 10.9% and accuracy ranged from 93.4 to 113.6% of nominal. Stable-labeled internal standard GDC-0980-d8 was used to minimize matrix effects. This assay was used for the measurement of GDC-0980 dog plasma concentrations to determine toxicokinetic parameters after oral administration of GDC-0980 (0.03, 0.1 and 0.3 mg/kg) to beagle dogs in a GLP toxicology study. Peak concentration ranged from 3.23 to 84.9 ng/mL. GDC-0980 was rapidly absorbed with a mean time to peak concentration ranging from 1.3 to 2.4 h. Mean area under the concentration-time curve from 0 to 24 hours ranged from 54.4 to 542 ng h/mL.