EMAIL THIS PAGE TO A FRIEND
Scientific Reports
S100A4 promotes pancreatic cancer progression through a dual signaling pathway mediated by Src and focal adhesion kinase.
PMID 25677816
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5585
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
C11H18ClN7O