EMAIL THIS PAGE TO A FRIEND
European Journal of Heart Failure
Rationale and design of MinerAlocorticoid Receptor antagonist Tolerability Study-Heart Failure (ARTS-HF): a randomized study of finerenone vs. eplerenone in patients who have worsening chronic heart failure with diabetes and/or chronic kidney disease.
PMID 25678098