EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Nearly 100% internal quantum efficiency in undoped electroluminescent devices employing pure organic emitters.
PMID 25678335
Abstract
The design of efficient and concentration-insensitive metal-free thermally activateddelayed fluorescence (TADF) materials is reported. Blue and green organic light-emitting diodes (OLEDs) containing a hole-transport layer, an undoped TADF emissive layer, and an electron-transport layer achieve maximum external quantum efficiencies of 19%, which is comparable to the best doped OLEDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900940
DMAC-DPS, sublimed
C42H36N2O2S