EMAIL THIS PAGE TO A FRIEND
Organic & Biomolecular Chemistry
Combining an Hsp70 inhibitor with either an N- or C-terminal Hsp90 inhibitor produces mechanistically distinct phenotypes.
PMID 25679754
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8476
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
C31H43N3O8