EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
The host plant metabolite glucose is the precursor of diffusible signal factor (DSF) family signals in Xanthomonas campestris.
PMID 25681189