EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Cutting edge: circulating plasmablasts induce the differentiation of human T follicular helper cells via IL-6 production.
PMID 25681343
Abstract
B cells require CD4(+) T follicular helper (Tfh) cells to progress through the germinal center and provide protective Ab responses. In this article, we reveal a reciprocal interaction whereby circulating human plasmablasts are potent inducers of the Tfh cell-differentiation program, including the expression of their key transcription factor Bcl-6. The markedly increased propensity of plasmablasts, compared with naive B cells, to induce Tfh cell differentiation was due to their increased production of IL-6. Specific targeting of IL-6 using tocilizumab therapy in patients with rheumatoid arthritis led to a significant reduction in circulating Tfh cell numbers and IL-21 production, which was correlated with reduced plasmablast formation. Our data uncover a positive-feedback loop between circulating plasmablasts and Tfh cells that could sustain autoimmunity and spread Ab-driven inflammation to unaffected sites; this represents an important therapeutic target, as well as reveals a novel mechanism of action for tocilizumab.