EMAIL THIS PAGE TO A FRIEND
Journal of Industrial Microbiology & Biotechnology
New lager yeast strains generated by interspecific hybridization.
PMID 25682107
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8378
Maltotriose, ≥90% (HPLC)
C18H32O16