EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology and hepatology
Dysfunction of liver regeneration in aged liver after partial hepatectomy.
PMID 25682855
Abstract
A remarkable feature of the liver is its regenerative capacity following partial hepatectomy. However, the regenerative capacity of many organs and tissues loses its natural ability to divide with aging. In this study, we investigated the association of aging with endoplasmic reticulum stress, the cell cycle, autophagy, and apoptosis-related genes during liver regeneration after hepatectomy. Balb/c 4-week and 40-week-old male mice were subjected to 70% hepatectomy. Animals were sacrificed at 24, 48, and 72 h after hepatectomy. Immunohistochemical stainings for proliferating cell nuclear antigen, LC3, Atg5, and caspase-3 were used to quantify protein expression. Real-time reverse transcription-polymerase chain reaction was used to detect p16, CHOP, LC3, Atg5, hepatocyte growth factor, cMet, cyclin D1, cyclin A2, and caspase-3 expression. After hepatectomy, old group showed a lower survival rate and significantly lower expression of hepatocyte growth factor, cMet, cyclin D1, cyclin A2, proliferating cell nuclear antigen labeling index, and SMP30 compared with young group. The liver weight/body weight ratio was significantly lower at 48 h and 72 h after hepatectomy and was accompanied by markedly elevated levels of the liver cell injury markers, LC3 and caspase-3. Immunohistochemical results showed that LC3, Atg5, and caspase-3 protein expression were higher in old group than in young group. These results revealed that impaired liver regeneration was due to aging, which was expressed by decreased cell cycle and increased autophagy and apoptosis. Therefore, understanding the molecular basis for aged liver regeneration might provide a new therapeutic option for old patients.