EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Quantitative analysis of mechanical and electrostatic properties of poly(lactic) acid fibers and poly(lactic) acid-carbon nanotube composites using atomic force microscopy.
PMID 25683087