EMAIL THIS PAGE TO A FRIEND
Steroids
Detection of dihydroxycholesterols in human plasma using HPLC-ESI-MS/MS.
PMID 25683891
Abstract
We report a straightforward sample preparation procedure and a direct liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) method for the analysis of 7alpha,25-dihydroxycholesterol (7α25-OHC) and 7alpha,27-dihydroxycholesterol (7α27-OHC). By applying a slow protein precipitation approach using cold ethanol, we were able to detect and quantify 7α25-OHC and 7α27-OHC in a fast and reliable manner. The average concentrations from 20 healthy individuals were determined to be 0.21±0.05nM for 7α25-OHC and 3.4±0.1nM for 7α27-OHC. In addition, we are the first to report the average degrees of esterification (n=8) to be 73.8% and 82% for 7α25-OHC and 7α27-OHC, respectively. Using the established method, we achieved the sensitivity sufficient for detecting low abundant dihydroxylated oxysterols in healthy individuals. This result should enable extension of these studies towards a comprehensive analysis of oxysterol levels under disease conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1015
25-Hydroxycholesterol, ≥98%
C27H46O2
SML0541
7α,25-Dihydroxycholesterol, ≥98% (HPLC)
C27H46O3