EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology and hepatology
Mediator subunit 23 overexpression as a novel target for suppressing proliferation and tumorigenesis in hepatocellular carcinoma.
PMID 25684393
Abstract
Hepatocellular carcinoma (HCC) is the fifth most frequent cancer in the world. However, the molecular mechanisms involved in HCC are still poorly understood. Here, we study the role of mediator subunit 23 (MED23), a component of the Mediator complex, in hepatocarcinogenesis. We detected MED23 expression in HCC samples by real-time polymerase chain reaction (PCR) and immunohistochemistry analysis. We also knocked down and overexpressed MED23 to explore its functional role in hepatoma cell growth. The cell cycle was examined by flow cytometry analysis, and protein expression was examined by Western blot. A xenograft mouse model was used to determine whether MED23 is involved in tumorigenesis. MED23 was frequently upregulated in human HCC tissues compared with paired adjacent non-tumorous liver tissues. The hepatoma cells also showed increased MED23 expression. MED23 knockdown inhibited hepatoma cell growth, whereas overexpression of MED23 promoted cell growth. Knockdown of MED23 induced a G1 to S phase arrest. Moreover, MED23 regulated the expression of p16(INK) (4a) (p16) and the phosphorylation of retinoblastoma protein (Rb). p16 was transcriptionally upregulated, and its promoter was demethylated. The G1 to S phase arrest induced by MED23 knockdown was aborted after p16 was silenced. Furthermore, MED23 knockdown suppressed tumorigenesis and regulated p16/Rb signaling in vivo. Taken together, our study suggests that MED23 plays an important role in hepatocarcinogenesis, and it may be a novel target for HCC therapy.