EMAIL THIS PAGE TO A FRIEND
World Journal of Gastroenterology
Human cytomegalovirus and Epstein-Barr virus infection in inflammatory bowel disease: need for mucosal viral load measurement.
PMID 25684960