EMAIL THIS PAGE TO A FRIEND
BioMed Research International
Prokineticin 2 upregulation in the peripheral nervous system has a major role in triggering and maintaining neuropathic pain in the chronic constriction injury model.
PMID 25685780