EMAIL THIS PAGE TO A FRIEND
Materials Science & Engineering. C, Materials For Biological Applications
Bioactivity of coatings formed on Ti-13Nb-13Zr alloy using plasma electrolytic oxidation.
PMID 25686936