EMAIL THIS PAGE TO A FRIEND
Archives of Virology
Baseline factors associated with treatment response in patients infected with hepatitis C virus 1b by stratification of IL28B polymorphisms.
PMID 25687192
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000377
Ribavirin, European Pharmacopoeia (EP) Reference Standard
C8H12N4O5