EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Loss of Nrf2 in bone marrow-derived macrophages impairs antigen-driven CD8(+) T cell function by limiting GSH and Cys availability.
PMID 25687825
Abstract
NF-E2-related factor 2 (Nrf2), known to protect against reactive oxygen species, has recently been reported to resolve acute inflammatory responses in activated macrophages. Consequently, disruption of Nrf2 promotes a proinflammatory macrophage phenotype. In the current study, we addressed the impact of this macrophage phenotype on CD8(+) T cell activation by using an antigen-driven coculture model consisting of Nrf2(-/-) and Nrf2(+/+) bone marrow-derived macrophages (BMDMΦ) and transgenic OT-1 CD8(+) T cells. OT-1 CD8(+) T cells encode a T cell receptor that specifically recognizes MHC class I-presented ovalbumin OVA(257-264) peptide, thereby causing a downstream T cell activation. Interestingly, coculture of OVA(257-264)-pulsed Nrf2(-/-) BMDMΦ with transgenic OT-1 CD8(+) T cells attenuated CD8(+) T cell activation, proliferation, and cytotoxic function. Since the provision of low-molecular-weight thiols such as glutathione (GSH) or cysteine (Cys) by macrophages limits antigen-driven CD8(+) T cell activation, we quantified the amounts of intracellular and extracellular GSH and Cys in both cocultures. Indeed, GSH levels were strongly decreased in Nrf2(-/-) cocultures compared to wild-type counterparts. Supplementation of thiols in Nrf2(-/-) cocultures via addition of glutathione ester, N-acetylcysteine, β-mercaptoethanol, or cysteine itself restored T cell proliferation as well as cytotoxicity by increasing intracellular GSH. Mechanistically, we identified two potential Nrf2-regulated genes involved in thiol synthesis in BMDMΦ: the cystine transporter subunit xCT and the modulatory subunit of the GSH-synthesizing enzyme γ-GCS (GCLM). Pharmacological inhibition of γ-GCS-dependent GSH synthesis as well as knockdown of the cystine antiporter xCT in Nrf2(+/+) BMDMΦ mimicked the effect of Nrf2(-/-) BMDMΦ on CD8(+) T cell function. Our findings demonstrate that reduced levels of GCLM as well as xCT in Nrf2(-/-) BMDMΦ limit GSH availability, thereby inhibiting antigen-induced CD8(+) T cell function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8630
DL-Cystine
C6H12N2O4S2
B10358
p-Benzoquinone, reagent grade, ≥98%
C6H4O2