PLoS ONE
OVCAR-3 spheroid-derived cells display distinct metabolic profiles.
PMID 25688563
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
45840
Acetic anhydride
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
386529
Chlorotrimethylsilane, purified by redistillation, ≥99%
C3H9ClSi
95541
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C3H9ClSi
92361
Chlorotrimethylsilane, ≥98.0% (GC)
C3H9ClSi
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
385433
Chlorotrimethylsilane solution, 1.0 M in methylene chloride
C3H9ClSi
384410
Chlorotrimethylsilane solution, 1.0 M in THF
C3H9ClSi
33045-U
Methoxyamine hydrochloride, bottle of 5 g
CH5NO · HCl
226904
Methoxyamine hydrochloride, 98%
CH5NO · HCl
225517
Methoxylamine hydrochloride solution, 25-30 wt. % in H2O
CH5NO · HCl
M0789
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥97.0% (GC)
C5H3F6NO2
M9513
N-Methyl-bis(trifluoroacetamide), ~95% (capillary GC)
C5H3F6NO2
65943
N-Methyl-bis(trifluoroacetamide), for GC derivatization, ≥99.0% (GC)
C5H3F6NO2