EMAIL THIS PAGE TO A FRIEND
Nucleic Acids Research
Single molecule analysis of Trypanosoma brucei DNA replication dynamics.
PMID 25690894
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6891
5-Chloro-2′-deoxyuridine, thymidine analog
C9H11ClN2O5