EMAIL THIS PAGE TO A FRIEND
Oncotarget
Myeloid differentiation primary response gene 88-leukotriene B4 receptor 2 cascade mediates lipopolysaccharide-potentiated invasiveness of breast cancer cells.
PMID 25691060
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
465119
Baicalein, 98%
C15H10O5