EMAIL THIS PAGE TO A FRIEND
Journal of Virology
Peptides designed to spatially depict the Epstein-Barr virus major virion glycoprotein gp350 neutralization epitope elicit antibodies that block virus-neutralizing antibody 72A1 interaction with the native gp350 molecule.
PMID 25694592