EMAIL THIS PAGE TO A FRIEND
Journal of Food Science and Technology
Palm-based medium-and-long-chain triacylglycerol (P-MLCT): production via enzymatic interesterification and optimization using response surface methodology (RSM).
PMID 25694677