EMAIL THIS PAGE TO A FRIEND
International Journal of Biological Macromolecules
Anti-cancer evaluation of quercetin embedded PLA nanoparticles synthesized by emulsified nanoprecipitation.
PMID 25701491