EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
RASSF4 promotes EV71 replication to accelerate the inhibition of the phosphorylation of AKT.
PMID 25701784
Abstract
Enterovirus 71 (EV71) is a neurotropic virus that causes hand, foot and mouth disease (HFMD), occasionally leading to death. As a member of the RAS association domain family (RASSFs), RASSF4 plays important roles in cell death, tumor development and signal transduction. However, little is known about the relationship between RASSF4 and EV71. Our study reveals for the first time that RASSF4 promotes EV71 replication and then accelerates AKT phosphorylation inhibition in EV71-infected 293T cells, suggesting that RASSF4 may be a potential new target for designing therapeutic measures to prevent and control EV71 infection.