EMAIL THIS PAGE TO A FRIEND
Talanta
Preparation of 4-butylaniline-bonded attapulgite for pre-concentration of bisphenol A in trace quantity.
PMID 25702981
Abstract
Ring-opening reaction with synchronous hydrolysis was used to prepare 4-butylaniline-modified attapulgite (abbreviated as BA-ATP) for pre-concentration of bisphenol A (BPA) in trace quantity. The preparation was achieved under mild condition, and the material was characterized by Fourier transform infrared spectroscopy (FT-IR), elemental analysis, nuclear magnetic resonance (NMR) and scanning electron microscopy. BA-ATP was used in solid phase extraction (SPE), and SPE was performed in a neutral condition without regard to ionic strength. The results indicate that BA-ATP has high affinity to BPA with a maximum adsorption amount of 44mg g(-1), and the adsorption can be described by Langmuir isotherm model. The content of bisphenol A in water samples was analyzed by HPLC method with the pre-concentration using BA-ATP. The limit of detection (LOD) can be as low as 3.9ng mL(-1), and the average recoveries are in a range of 93-97% with relative standard deviation (RSD) of less than 2%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112666
4-Butylaniline, 97%
C10H15N