EMAIL THIS PAGE TO A FRIEND
Talanta
Systematic evaluation of mobile phase additives for the LC-MS characterization of therapeutic proteins.
PMID 25702986
Abstract
Trifluoroacetic acid (TFA) is commonly used as mobile phase additive for the analysis of proteins in reversed phase liquid chromatography (RPLC). Due to its interesting features, it provides symmetrical and narrow peak shapes for proteins, but decreases mass spectrometric sensitivity through ion-pairing and spray destabilizing. Since RPLC-MS is an important technique for the characterization of proteins, some alternative MS-compatible mobile phases may be required. The aim of this study was to evaluate various acidic and basic mobile phase additives for the LC-MS analysis of therapeutic proteins possessing molecular weight between 5 and 150kDa. At the end, 10mM formate buffer pH 3 was found to be the most promising alternative, since it provided acceptable peak shapes in most cases, together with an average improvement of MS sensitivity by 5-times, compared to TFA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52517
1,1,1,3,3,3-Hexafluoro-2-propanol, for GC derivatization, LiChropur, ≥99.8%
C3H2F6O