EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Gas chromatography-mass spectrometry method optimized using response surface modeling for the quantitation of fungal off-flavors in grapes and wine.
PMID 25703150
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74858
(±)-2-Octanol, ReagentPlus®, ≥99.5% (GC)
C8H18O
42277
(±)-2-Octanol, analytical standard
C8H18O
74860
(±)-2-Octanol, ≥96.0% (GC)
C8H18O
O5284
1-Octen-3-ol, 98%
C8H16O
W280518
1-Octen-3-ol, ≥98%, FCC, FG
C8H16O
W280523
1-Octen-3-ol, natural, ≥95%, FG
C8H16O
777064
1-Octen-3-one, 96%
C8H14O
398780
2,4,6-Tribromoanisole, 99%
C7H5Br3O
655147
2,4,6-Trichloroanisole-d5, 98 atom % D
C7D5Cl3O
34023
2,4,6-Trichloroanisole-d5, PESTANAL®, analytical standard
C7D5Cl3O
243132
2-Isopropyl-3-methoxypyrazine, 97%
C8H12N2O
W335808
2-Isopropyl-3-methoxypyrazine, 99%, FG
C8H12N2O
743364
2-Methylisoborneol, ≥98.0% (GC)
C11H20O
M3933
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)
C11H20O
details
2-Methylisoborneol solution, analytical standard
C11H20O
CRM47523
2-Methylisoborneol solution, certified reference material, TraceCERT®, 100 μg/mL in methanol, ampule of 1 mL
C11H20O
O4504
2-Octanol, 97%
C8H18O
W280100
2-Octanol, ≥97%
C8H18O
W280110
2-Octanol, natural, ≥97%, FG
C8H18O
46214
3-Octanone, analytical standard
C8H16O
136913
3-Octanone, ≥98%
C8H16O
W280305
3-Octanone, ≥98%, FG
C8H16O
293261
4-Methyl-2-pentanone, for HPLC, ≥99.5%
C6H12O
360511
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
C6H12O
02474
4-Methyl-2-pentanone, analytical standard
C6H12O
W273104
4-Methyl-2-pentanone, ≥99%, FCC
C6H12O
537713
4-Methyl-2-pentanone, ≥99%
C6H12O
I13901
Isoborneol, 95%
C10H18O
W215805
Isoborneol, ≥95%, FG
C10H18O
251380
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
C4H6O6