EMAIL THIS PAGE TO A FRIEND
Acta medica Okayama
ONO-1301, a sustained-release prostacyclin analog, ameliorates the renal alterations in a mouse type 2 diabetes model possibly through its protective effects on mesangial cells.
PMID 25703166
Abstract
Diabetic nephropathy is the most common pathological disorder predisposing patients to end-stage renal disease. Considering the increasing prevalence of type 2 diabetes mellitus worldwide, novel therapeutic approaches are urgently needed. ONO-1301 is a novel sustained-release prostacyclin analog that inhibits thromboxane A2 synthase. Here we examined the therapeutic effects of the intermittent administration of slow-release ONO-1301 (SR-ONO) on diabetic nephropathy in obese type 2 diabetes mice, as well as its direct effects on mesangial cells. The subcutaneous injection of SR-ONO (3mg/kg) every 3 wks did not affect the obesity or hyperglycemia in the db/db obese mice used as a model of type 2 diabetes, but it significantly ameliorated their albuminuria, glomerular hypertrophy, glomerular accumulation of type IV collagen, and monocyte/macrophage infiltration, and also the increase of TGF-β1, α-smooth muscle actin (α-SMA) and MCP-1 compared to vehicle treatment. In cultured mouse mesangial cells, ONO-1301 concentration-dependently suppressed the increases in TGF-β, type IV collagen, α-SMA, MCP-1 and fibronectin induced by high ambient glucose, at least partly through prostacyclin (PGI2) receptor-mediated signaling. Taken together, these results suggest the potential therapeutic efficacy of the intermittent administration of SR-ONO against type 2 diabetic nephropathy, possibly through protective effects on mesangial cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2264
ONO-1301, ≥98% (HPLC)
C26H24N2O4