EMAIL THIS PAGE TO A FRIEND
Toxicology Letters
Indoxyl sulfate induces oxidative stress and hypertrophy in cardiomyocytes by inhibiting the AMPK/UCP2 signaling pathway.
PMID 25703824
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3490
Ponasterone A, ≥65%
C27H44O6